Learn More
Invitrogen™ PTCH Polyclonal Antibody

Description
PA1-4134 detects Patched (PTCH) in human samples. PA1-4134 has been successfully used in Western blot procedures. By Western blot this antibody detects a ~158 kDa protein representing PTCH in fetal brain lysate. The PA1-4134 immunogen is a synthetic peptide corresponding to residues K(269) K I N Y Q V D S W E(279) of mouse PTCH. This sequence is 100% conserved in human, rat, and chicken PTCH, and is expected to cross react with these samples.
Specifications
Specifications
| Antigen | PTCH |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | Ptch1 |
| Gene Accession No. | Q13635, Q61115, Q90693 |
| Gene Alias | A230106A15Rik; BCNS; etID309849.2; FLJ26746; FLJ42602; HGNC:9585; HPE7; lep; leprechaun; mes; mesenchymal dysplasia; NBCCS; OTTHUMP00000021709; OTTHUMP00000021710; patched; Patched 1; patched 2; patched homolog 1; patched homolog 2; patched protein; patched1; patched-2 protein; patched-like protein; protein patched homolog 1; Ptc; ptc1; ptc-1; ptc2; Ptch; PTCH protein +12b; PTCH protein +4'; PTCH protein -10; PTCH protein -3; PTCH protein -3,4,5; PTCH1; Ptch1 protein +12b; PTCH11; Ptch2; RP11-435O5.3 |
| Gene Symbols | Ptch1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.