Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PTCH Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110235
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110235 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110235 Supplier Thermo Scientific Supplier No. PA5110235
Add to Cart
Add to Cart

PTCH Polyclonal Antibody

PTCH Polyclonal Antibody for Western Blot, ICC/IF

TRUSTED_SUSTAINABILITY

Specifications

Antigen PTCH
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 2.77 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Ptch1
Gene Accession No. Q13635, Q61115
Gene Alias A230106A15Rik; BCNS; etID309849.2; FLJ26746; FLJ42602; HGNC:9585; HPE7; lep; leprechaun; mes; mesenchymal dysplasia; NBCCS; OTTHUMP00000021709; OTTHUMP00000021710; patched; Patched 1; patched 2; patched homolog 1; patched homolog 2; patched protein; patched1; patched-2 protein; patched-like protein; protein patched homolog 1; Ptc; ptc1; ptc-1; ptc2; Ptch; PTCH protein +12b; PTCH protein +4'; PTCH protein -10; PTCH protein -3; PTCH protein -3,4,5; PTCH1; Ptch1 protein +12b; PTCH11; Ptch2; RP11-435O5.3
Gene Symbols Ptch1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 770-970 of human PTCH1 (NP_000255.2).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19206, 5727, 89830
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.