Learn More
PTCH2 Mouse anti-Human, Clone: OTI1F5, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF804800

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Mutations of the human Patched tumor suppressor gene (PTCH) have been identified in individuals with the nevoid basal cell carcinoma syndrome (NBCCS) as well as in sporadic basal cell carcinomas and medulloblastomas. PTCH2 has a frameshift mutation resulting in a truncated PTCH2 protein.Specifications
PTCH2 | |
Monoclonal | |
Unconjugated | |
PTCH2 | |
PTC2 | |
Mouse | |
Affinity Chromatography | |
RUO | |
8643 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI1F5 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q9Y6C5 | |
PTCH2 | |
Human recombit protein fragment corresponding to amino acids 79-392 of human PTCH2 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.