Learn More
Invitrogen™ PTCH2 Polyclonal Antibody

Description
PA1-4135 detects Patched 2 (PTCH2) in human and mouse samples. PA1-4135 has been successfully used in Western blot procedures. By Western blot this antibody detects a ~158 kDa protein representing PTCH2 in fetal brain lysate. The PA1-4135 immunogen is a synthetic peptide corresponding to residues G(226) P F A S L E G F R(235) of human PTCH2.
Specifications
Specifications
| Antigen | PTCH2 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | PTCH2 |
| Gene Accession No. | O35595, Q9Y6C5 |
| Gene Alias | patched 2; patched homolog 1; patched homolog 2; protein patched homolog 2; PTC2; Ptch1; PTCH2; RP11-269F19.8; UNQ560/PRO1121/PRO57079 |
| Gene Symbols | PTCH2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.