Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PTCH2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113810
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113810 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113810 Supplier Invitrogen™ Supplier No. PA5113810
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LAQEALPENA SQQIHAFSST TLDDILHAFS EVSAARVVGG YLLMLAYACV For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Dog: 100%; Guinea Pig: 79%; Horse: 100%; Human: 100%; Mouse: 93%; Rabbit: 86%; Rat: 93%.

Mutations of the human Patched tumor suppressor gene (PTCH) have been identified in individuals with the nevoid basal cell carcinoma syndrome (NBCCS) as well as in sporadic basal cell carcinomas and medulloblastomas. PTCH2 has a frameshift mutation resulting in a truncated PTCH2 protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PTCH2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene PTCH2
Gene Accession No. Q9Y6C5
Gene Alias patched 2; patched homolog 1; patched homolog 2; protein patched homolog 2; PTC2; Ptch1; PTCH2; RP11-269F19.8; UNQ560/PRO1121/PRO57079
Gene Symbols PTCH2
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N terminal region of human PTCH2 (aa 362-411).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8643
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.