Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PTCHD1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA548191
Description
PTCHD1 Polyclonal Antibody for Western Blot, IHC (P)
This gene encodes a membrane protein with a patched domain. The encoded protein is similar to Drosophila proteins which act as receptors for the morphogen sonic hedgehog. Deletions in this gene, which is located on the X chromosome, are associated with intellectual disability and autism (PMID: 21091464, PMID: 20844286). [provided by RefSeq, Aug 2011].
Specifications
PTCHD1 | |
Polyclonal | |
Unconjugated | |
PTCHD1 | |
9630036J22Rik; AI848149; AUTSX4; FLJ30296; Gm387; MGC149798; patched domain containing 1; patched domain-containing protein 1; PTCHD1; PTCHD1 isoform | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
139411, 211612 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.25 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
Q14B62, Q96NR3 | |
PTCHD1 | |
KLH conjugated synthetic peptide between 611-639 amino acids from the Central region of human PTCHD1. | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction