Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PTCHD2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562984
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562984 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562984 Supplier Invitrogen™ Supplier No. PA562984
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PSGYSISSFL QMLHPECKEL PEPNLLPGQL SHGAVGVREG RVQWISMAFE STTYKGKSSF QTYSDYLRWE SFLQQQ Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 97%.

Plays a role in neuronal proliferation and differentiation (PubMed:25281927). Plays a role in the accumulation of cellular cholesterol. Involved in intracellular lipid droplet formation (PubMed:25281927). May contribute to cholesterol homeostasis in neuronal cells. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen PTCHD2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DISP3
Gene Accession No. Q9P2K9
Gene Alias DISP3; dispatched 3; dispatched homolog 3; dispatched RND transporter family member 3; G630052C06Rik; Kiaa1337; patched domain containing 2; patched domain-containing protein 2; Protein dispatched homolog 3; Ptchd2; RGD1305820; RNDEu-2; RND-type protein RNDEu-2
Gene Symbols DISP3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PTCHD2. Recombinant protein control fragment (Product #RP-101619).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57540
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.