Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PTCHD2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA572748
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA572748 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA572748 Supplier Invitrogen™ Supplier No. PA572748
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PTCHD2 Polyclonal Antibody for Western Blot, IHC (P), ELISA

Plays a role in neuronal proliferation and differentiation (PubMed:25281927). Plays a role in the accumulation of cellular cholesterol. Involved in intracellular lipid droplet formation (PubMed:25281927). May contribute to cholesterol homeostasis in neuronal cells. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen PTCHD2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene DISP3
Gene Accession No. A3KFU9, Q9P2K9
Gene Alias DISP3; dispatched 3; dispatched homolog 3; dispatched RND transporter family member 3; G630052C06Rik; Kiaa1337; patched domain containing 2; patched domain-containing protein 2; Protein dispatched homolog 3; Ptchd2; RGD1305820; RNDEu-2; RND-type protein RNDEu-2
Gene Symbols DISP3
Host Species Rabbit
Immunogen PTCHD2 antibody for a 19 amino acid synthetic peptide from near the center of human PTCHD2. The immunogen is within amino acids 780 - 830 of PTCHD2.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 242748, 313705, 57540
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.