Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PTCHD3 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982142
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2142 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2142 Supplier Bioss Supplier No. BS12325R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

May play a role in sperm development or sperm function.

Specifications

Antigen PTCHD3
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene PTCHD3
Gene Accession No. Q3KNS1
Gene Alias FLJ44037; MGC129888; patched domain containing 3; Patched domain-containing protein 3; Patched-related protein; PTCHD3; PTR
Gene Symbols PTCHD3
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human PTCHD3.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 374308
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.