Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PTCHD4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5118217
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5118217 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5118217 Supplier Invitrogen™ Supplier No. PA5118217
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Specificity: Human PTCHD4.

TRUSTED_SUSTAINABILITY

Specifications

Antigen PTCHD4
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.03% ProClin 300; pH 7
Gene PTCHD4
Gene Accession No. Q6ZW05
Gene Alias 3110082D06Rik; C6orf138; dJ402H5.2; Gm321; p53-regulated patched protein; patched domain containing 4; patched domain-containing protein 4; patched domain-containing protein C6orf138; patched domain-containing protein C6orf138 homolog; PTCH53; PTCHD4; RGD1563020
Gene Symbols PTCHD4
Host Species Rabbit
Immunogen E. coli-derived Human PTCHD4 fragment.
Purification Method Antigen Affinity Chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 442213
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.