Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PTCRA Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA549122
Description
PTCRA Polyclonal Antibody for Western Blot
The pre-T-cell receptor complex (composed of PTCRA, TCRB and the CD3 complex) regulates early T-cell development.
Specifications
PTCRA | |
Polyclonal | |
Unconjugated | |
PTCRA | |
pre T-cell antigen receptor alpha; pre-T-cell receptor alpha chain; PTA; pT-alpha; pT-alpha-TCR; PTCRA | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
171558 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.45 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
Q6ISU1 | |
PTCRA | |
KLH conjugated synthetic peptide between 197-225 amino acids from the C-terminal region of human PTCRA. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction