Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PTDSS1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5112828
Description
PTDSS1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA
Catalyzes a base-exchange reaction in which the polar head group of phosphatidylethanolamine (PE) or phosphatidylcholine (PC) is replaced by L-serine. In membranes, PTDSS1 catalyzes mainly the conversion of phosphatidylcholine. Also converts, in vitro and to a lesser extent, phosphatidylethanolamine.
Specifications
| PTDSS1 | |
| Polyclonal | |
| Unconjugated | |
| PTDSS1 | |
| KIAA0024; LMHD; Phosphatidylserine synthase 1; PSS1; PSS-1; PSSA; ptdSer synthase 1; PTDSS1; Serine-exchange enzyme I | |
| Rabbit | |
| Protein G | |
| RUO | |
| 9791 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| P48651 | |
| PTDSS1 | |
| Recombinant Human Phosphatidylserine synthase 1 protein (1-35AA). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction