Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ PTEN-alpha, Mouse, Unlabeled, Clone: 3A4.1,

Catalog No. MABS1680MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
MABS1680MI 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABS1680MI Supplier MilliporeSigma™ Supplier No. MABS1680
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Phosphatidylinositol 3,4,5-trisphosphate 3-phosphatase and dual-specificity protein phosphatase PTEN (EC 3.1.3.16, EC 3.1.3.48, EC 3.1.3.67; UniProt P60484; also known as Mitochondrial PTENalpha, Phosphatase and tensin-like protein, Mutated in multiple advanced cancers 1, Mitochondrial phosphatase and tensin protein alpha, phosphatase and tensin homolog deleted on chromosome 10) is encoded by the PTEN (also known as 10q23del, BRRS, BZS, CWS1, GLM2, MMAC1, MHAM, TEP1) gene (Gene ID 5728) in human. PTEN is a tumor suppressor that catalyzes the removal of phosphate from phosphoserine, phosphothreonine, and phosphotyrosine residues on substrate proteins, as well as the phosphate in the D3 position of the inositol ring from phosphatidylinositol 3,4,5-trisphosphate, phosphatidylinositol 3,4-diphosphate, phosphatidylinositol 3-phosphate and inositol 1,3,4,5-tetrakisphosphate. PTEN plays a major role in controlling cell growth and proliferation, and PTEN gene mutains are found in a large number of cancers at high frequency. PTEN activity, stability, localization, and conformation are controlled through complex regulatory mechanisms, including the reversible phosphorylations of several Ser/Thr (S/T) residues in its unfolded C-terminal tail region (PTEN-Ctail, residues 351 403). Alternative translation start site 519 base pairs upstream of the ATG initiation sequence results in the translational variant of PTEN-alpha (PTEN-Long), including additional 173 N-terminal amino acids to the canonical 403-a.a. PTEN sequence. PTEN-Long is a membrane-permeable phosphatase secreted by PTEN-Long producing cells and can enter other cells as an exogenous PI3K signaling-inhibitory agent. PTEN-Long is reported to induce tumor cell death in vitro and in vivo.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PTEN-alpha
Applications Western Blot
Classification Monoclonal
Clone 3A4.1
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified mouse monoclonal antibody IgG2aκ in buffer containing 0.1M Tris-Glycine (pH 7.4) 150mM NaCl with 0.05% Sodium Azide.
Gene PTEN, 10q23del, BRRS, BZS, CWS1, GLM2, MMAC1, MHAM, TEP1
Gene Accession No. P60484-2
Host Species Mouse
Immunogen GST-tagged recombinant human PTEN-alpha N-terminal fragment.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Gene ID (Entrez) NP_001291646
Test Specificity Clone 3A4.1 targets an N-terminal region specific to the spliced isoform alpha (PTEN-Long; P60484-2) and not present in spliced isoform 1 (a.k.a. 55 kDa isoform) or isoform 3.
Target Species Human
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.