Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PTEN Antibody (1042824), Alexa Fluor™ 700, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB6655N
Description
PTEN Monoclonal antibody specifically detects PTEN in Human samples. It is validated for Flow CytometrySpecifications
PTEN | |
Monoclonal | |
Alexa Fluor 700 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
1042824 | |
Intracellular Staining by Flow Cytometry | |
BZS, EC 3.1.3.16,10q23del, EC 3.1.3.48, EC 3.1.3.67, GLM2, MMAC1 phosphatase and tensin homolog deleted on chromosome 10, MMAC1MGC11227, Mutated in multiple advanced cancers 1, phosphatase and tensin homologDEC, phosphatidylinositol-34,5-trisphosphate 3-phosphatase and dual-specificityprotein phosphatase PTEN, PTEN1, TEP1MHAM | |
E. coli-derived recombinant human PTEN, Thr2-Val403, Accession # P60484 | |
0.1 mL | |
Autophagy, Cancer, Cell Biology, Lipid and Metabolism, Metastasis, mTOR Pathway, Phospho Specific, Protein Phosphatase, Signal Transduction, Transcription Factors and Regulators, Tumor Suppressors | |
5728 | |
Store at 4C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction