Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PTEN Antibody (CL11722), Novus Biologicals™

Mouse Monoclonal Antibody
$340.50 - $557.50
Specifications
Antigen | PTEN |
---|---|
Clone | CL11722 |
Dilution | Western Blot 1 ug/ml, Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000 |
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Description
PTEN Monoclonal antibody specifically detects PTEN in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
PTEN | |
Western Blot 1 ug/ml, Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
BZS, EC 3.1.3.16,10q23del, EC 3.1.3.48, EC 3.1.3.67, GLM2, MMAC1 phosphatase and tensin homolog deleted on chromosome 10, MMAC1MGC11227, Mutated in multiple advanced cancers 1, phosphatase and tensin homologDEC, phosphatidylinositol-34,5-trisphosphate 3-phosphatase and dual-specificityprotein phosphatase PTEN, PTEN1, TEP1MHAM | |
This antibody was developed against a Synthetic Peptide of P60484. Actual immunogen sequence is proprietary. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
CL11722 | |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Autophagy, Cancer, Cell Biology, Lipid and Metabolism, Metastasis, mTOR Pathway, Phospho Specific, Protein Phosphatase, Signal Transduction, Transcription Factors and Regulators, Tumor Suppressors | |
PBS, pH 7.2, 40% glycerol | |
5728 | |
IgG1 | |
Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title