Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PTEN-Long Mouse anti-Human, Clone: 1C2, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MABS450B25UG
Description
PTEN-Long Monoclonal antibody specifically detects PTEN-Long in Human samples. It is validated for ELISA, Western Blotting.
Specifications
PTEN-Long | |
Monoclonal | |
1.0 mg/mL | |
Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide. | |
A bacterial fusion protein with a V5/His-tag at its C- terminus. | |
25 μg | |
Primary | |
Stable for 1 year at +2°C to +8°C from date of receipt. |
ELISA, Western Blot | |
1C2 | |
Unconjugated | |
Mouse | |
Protein G purified | |
RUO | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction