Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PTEN mono-ubiquitylated K13 Rabbit anti-Human, Unconjugated, Polyclonal, MilliporeSigma™

Catalog No. ABS53325UL Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABS53325UL 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABS53325UL Supplier MilliporeSigma Supplier No. ABS53325UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-PTEN mono-ubiquitylated K13, Cat. No. ABS533, is a rabbit polyclonal antibody that detects phosphatidylinositol 3,4,5-trisphosphate 3-phosphatase and dual-specificity protein phosphatase PTEN and has been tested for use in Western Blotting and Peptide Inhibition Assay.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PTEN mono-ubiquitylated K13
Applications Western Blot, Inhibition Assays
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias Phosphatidylinositol 3;4;5-trisphosphate 3-phosphatase and dual-specificity protein phosphatase PTEN;Mutated in multiple advanced cancers 1;Phosphatase and tensin homolog;Phosphatase and tensin homologue deleted on chromosome 10
Gene Symbols PTEN;MMAC1;TEP1
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to 12 amino acids surrounding ubiquitinated Lysine 13 from the N-terminal region of human PTEN.
Molecular Weight of Antigen ∽65 kDa observed; 47.17 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
Purification Method Affinity Purified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Test Specificity This rabbit polyclonal antibody detects phosphatidylinositol 3,4,5-trisphosphate 3-phosphatase and dual-specificity protein phosphatase PTEN monoubiquitinated on Lysine 13. Immunogen sequence is conserved, hence, it is expected to react with all 3 isoforms.
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Product Type Antibodies
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.