Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PTEN2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562027
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562027 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562027 Supplier Invitrogen™ Supplier No. PA562027
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MNESPDPTDL AGVIIELGPN DSPQT Highest antigen sequence identity to the following orthologs: Mouse - 40%, Rat - 40%.

TPTE encodes a putative transmembrane tyrosine phosphatase that may be involved in signal transduction pathways of the endocrine or spermatogenetic function of the testis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PTEN2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TPTE
Gene Accession No. P56180
Gene Alias Cancer/testis antigen 44; CT44; Pten2; PTEN-related tyrosine phosphatase; Putative tyrosine-protein phosphatase TPTE; tensin, putative protein-tyrosine phosphatase; TPTE; Transmembrane phosphatase with tensin homology; tumor antigen BJ-HCC-5
Gene Symbols TPTE
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PTEN2. Recombinant protein control fragment (Product #RP-95900).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7179
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.