Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PTENP1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB11086973
Description
PTENP1 Polyclonal specifically detects PTENP1 in Human samples. It is validated for Western Blot, ELISA.Specifications
PTENP1 | |
Polyclonal | |
Unconjugated | |
0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide | |
phosphatase and tensin homolog pseudogene 1, psiPTEN, PTEN2, PTH2, PTEN-rs | |
Rabbit | |
47.16 kDa | |
0.1 mg | |
Protein Phosphatase | |
11191 | |
Human, Mouse, Mammalia, Canine, Monkey, Opossum, Pufferfishes, Primate, Zebrafish | |
IgG |
Western Blot, ELISA | |
1.15 mg/mL | |
Western Blot 1:500-1:2000, ELISA 1:160000 | |
O43460 | |
PTENP1 | |
This affinity purified PTENP1 Antibody was prepared from whole rabbit serum produced by repeated immunizations with a peptide corresponding to amino acids near the N-terminal end of human PTENP1 protein. (Uniprot: O43460) | |
Affinity Purified | |
RUO | |
Primary | |
This is directed against human PTEN-P1 protein. | |
Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction