Learn More
3B Scientific™ Pteriodophytes (Ferns and Fern Allies) Slide Set
SDPSCI_ED
Catalog No. s95053l
Click to view available options
Quantity:
15
15 microscope slides
Primitive ferns (Psilophytatae):
  • 1(d). Psilotum, rhizome showing protostele t.s.
  • 2(d). Psilotum, stem showing squamous leaves, aktinostele t.s. Clubmosses (Lycopodiatae)
  • 3(c). Lycopodium, of stem showing plectostele t.s.
  • 4(e). Lycopodium, of strobilus showing isospores t.s.
  • 5(c). Selaginella, stem showing siphonostele t.s. Horse-tails (Equisetatae)
  • 6(c). Equisetum, stem t.s.
  • 7(d). Equisetum, strobilus showing spores t.s.
  • 8(b). Equisetum, w.m. of spores with elaters Ferns (Filicatae)
  • 9(c). Aspidium, root t.s.
  • 10(c). Aspidium, stem t.s.
  • 11(d). Aspidium, leaf showing sori t.s.
  • 12(f). Aspidium, w.m. of prothallium showing antheridia and archegonia
  • 13(d). Pteridium, rhizome t.s.
  • 14(d). Osmunda, royal fern, rhizome with ectophloic siphonostele t.s.
  • 15(d). Phyllitis scolopendrium, hart's tongue fern, leaf with sori and sporangia t.s.
Specifications
Type Ferns/Allies
Quantity 15
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.