Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PTF1A Antibody, Biotin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298726B
Description
PTF1A Polyclonal specifically detects PTF1A in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
PTF1A | |
Polyclonal | |
PBS | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Immunofluorescence | |
Biotin | |
bHLH transcription factor p48, BHLHA29, bHLHa29exocrine pancreas-specific transcription factor p48, Class A basic helix-loop-helix protein 29, p48 DNA-binding subunit of transcription factor PTF1, pancreas specific transcription factor, 1a, pancreas transcription factor 1 subunit alpha, Pancreas-specific transcription factor 1a, PTF1P48, PTF1-p48class II bHLH protein PTF1A | |
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of human PTF1A. | |
0.1 mL | |
Protein Kinase | |
256297 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction