Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PTF1A Antibody, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NB10093539
Description
PTF1A Polyclonal specifically detects PTF1A in Zebrafish samples. It is validated for Peptide ELISA.Specifications
PTF1A | |
Polyclonal | |
Unconjugated | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
bHLH transcription factor p48, BHLHA29, bHLHa29exocrine pancreas-specific transcription factor p48, Class A basic helix-loop-helix protein 29, p48 DNA-binding subunit of transcription factor PTF1, pancreas specific transcription factor, 1a, pancreas transcription factor 1 subunit alpha, Pancreas-specific transcription factor 1a, PTF1P48, PTF1-p48class II bHLH protein PTF1A | |
Goat | |
Affinity Purified | |
RUO | |
Primary | |
Zebrafish | |
IgG |
ELISA | |
0.5 mg/mL | |
Peptide ELISA Detection limit 1:16000 | |
NP_997524.1 | |
PTF1A | |
Peptide with sequence C-EDPRKLHLKSSINN corresponding to internal region according to Uniprot Zebrafish NP_997524.1. | |
0.1 mg | |
Protein Kinase | |
256297 | |
Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction