Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PTF1A Monoclonal Antibody (OTI5E5), OriGene
SDP

Catalog No. 502543993
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-3993 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-254-3993 Supplier OriGene Supplier No. CF810945
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PTF1A Monoclonal antibody specifically detects PTF1A in Human samples. It is validated for Western Blot

Specifications

Antigen PTF1A
Applications Western Blot
Classification Monoclonal
Clone OTI5E5
Concentration See Label
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene HOXD8
Gene Accession No. P13378
Gene Alias bHLH transcription factor; bHLH transcription factor p48; BHLHA29; class A basic helix-loop-helix protein 29; class II bHLH protein PTF1A; exocrine pancreas-specific transcription factor p48; p48 DNA-binding subunit of transcription factor PTF1; PACA; PAGEN2; pancreas associated transcription factor 1a; pancreas specific transcription factor, 1a; pancreas transcription factor 1 subunit alpha; pancreas transcription factor1 p48 subunit; pancreas-specific transcription factor 1a; PTF1; PTF1A; PTF1p48; PTF1-p48; Ptfa; si:zc142h2.2; zgc:112216
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 69-290 of human HOXD8 produced in E.coli.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3234
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Lyophilized
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.