Learn More
Synthware™ PTFE Protected High Vacuum Valves
GREENER_CHOICESDP
Catalog No. 31502336
Click to view available options
Size:
0 to 12 mm
0 to 4 mm
0 to 8 mm
High vacuum valves are fully enclosed, and include teflon-covered O-rings for increased durability.
  • PTFE protected high vacuum valves
  • Teflon-covered O-rings offer increased protection
  • Fully enclosed
Specifications
Description Fully Enclosed
For Use With (Application) Vacuum Sealing
Includes Teflon Covered O-Rings
Material PTFE
Type Valve
Size 0 to 12 mm
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.