Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BUCHI PTFE Tubing

For use with industrial Rotavapor Rotary Evaporators
$19.50 - $52.77
Specifications
Color | White |
---|---|
Material | PTFE |
For Use With (Application) | Introducing Solvent, Back Feeding |
Catalog Number | Mfr. No. | Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Type | Price | Quantity & Availability | |||||
05-405-020
![]() |
BUCHI
028096 |
Feeding Hose |
Each for $19.50
|
|
|||||
05-405-019
![]() |
BUCHI
027277 |
Vacuum Hose |
Each for $52.77
|
|
|||||
Description
- PTFE Tubing
- For use with Rotavapor Rotary Evaporators
Specifications
White | |
PTFE | |
Introducing Solvent, Back Feeding |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title