Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PTGDR Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA534114
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA534114 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA534114 Supplier Invitrogen™ Supplier No. PA534114
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon (100%) Monkey (90%) Marmoset (85%).

The protein encoded by this gene is a G-protein-coupled receptor. It has been shown to function as a prostanoid DP receptor. The activity of this receptor is mainly mediated by G-S proteins that stimulate adenylate cyclase resulting in an elevation of intracellular cAMP and Ca2+. Knockout studies in mice suggest that the ligand of this receptor, prostaglandin D2 (PGD2), functions as a mast cell-derived mediator to trigger asthmatic responses.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PTGDR
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene PTGDR
Gene Accession No. Q13258
Gene Alias AS1; ASRT1; DP; Dp receptor; DP1; MGC49004; PGD receptor; PGD2 receptor; prostaglandin D2 receptor; prostaglandin D2 receptor (DP); Prostanoid DP receptor; PTGDR; PTGDR1
Gene Symbols PTGDR
Host Species Rabbit
Immunogen Synthetic 20 amino acid peptide from N-terminal extracellular domain of human Prostaglandin D2 Receptor.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5729
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.