Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PTGDS Monoclonal Antibody (07)
GREENER_CHOICE

Catalog No. PIMA530796
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA530796 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA530796 Supplier Invitrogen™ Supplier No. MA530796
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human PTGDS/L-PGDS.

The protein encoded by this gene is a glutathione-independent prostaglandin D synthase that catalyzes the conversion of prostaglandin H2 (PGH2) to postaglandin D2 (PGD2). PGD2 functions as a neuromodulator as well as a trophic factor in the central nervous system. PGD2 is also involved in smooth muscle contraction/relaxation and is a potent inhibitor of platelet aggregation. PTGDS has also been implicated in the development and maintenance of the blood-brain, blood-retina, blood-aqueous humor and blood-testis barrier. Studies with transgenic mice overexpressing this gene suggest that this gene may be also involved in the regulation of non-rapid eye movement (NREM) sleep. It is likely to play important roles in both maturation and maintenance of the central nervous system and male reproductive system. PTGDS is the most abundant protein in the cerebral spinal fluid and recent evidence suggests that PTGDS acts as a Beta-Amyloid chaperone and may have a role in the deposition of Ab plaques in Alzheimer's disease.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PTGDS
Applications ELISA
Classification Monoclonal
Clone 07
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene PTGDS
Gene Accession No. P41222
Gene Alias 21kDa; Beta-trace protein; cerebrin-28; Glutathione-independent PGD synthase; glutathione-independent PGD synthetase; lipocalin-type prostaglandin D synthase; lipocalin-type prostaglandin-D synthase; LPGDS; L-PGDS; PDS; PGD2; PGD2 synthase; PGDS; PGDS2; PH2DISO; Prostaglandin D synthase; prostaglandin D2 synthase; prostaglandin D2 synthase (21 kDa, brain); prostaglandin D2 synthase (brain); prostaglandin D2 synthase 21kDa (brain); prostaglandin-D2 synthase; Prostaglandin-H2 D-isomerase; Ptgds; Ptgs3; testis tissue sperm-binding protein Li 63n
Gene Symbols PTGDS
Host Species Mouse
Immunogen Recombinant Human PTGDS/L-PGDS Protein (Met1-Gln190).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5730
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.