Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PTGER1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA534118
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA534118 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA534118 Supplier Invitrogen™ Supplier No. PA534118
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Human, Gorilla, Monkey, Marmoset, Hamster, Bovine, Dog (94%) Mouse, Rat, Bat (81%).

The protein encoded by this gene is a member of the G protein-coupled receptor family. This protein is one of four receptors identified for prostaglandin E2 (PGE2). Through a phosphatidylinositol-calcium second messenger system, G-Q proteins mediate this receptor's activity. Knockout studies in mice suggested a role of this receptor in mediating algesia and in regulation of blood pressure. Studies in mice also suggested that this gene may mediate adrenocorticotropic hormone response to bacterial endotoxin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PTGER1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene Ptger1
Gene Accession No. P34995
Gene Alias 42kDa; EP1; EP1 prostanoid receptor; PGE receptor EP1 subtype; PGE receptor, EP1 subtype; PGE2 receptor EP1 subtype; prostaglandin E receptor 1; prostaglandin E receptor 1 (subtype EP1); prostaglandin E receptor 1 (subtype EP1), 42kD; prostaglandin E receptor 1 (subtype EP1), 42kDa; prostaglandin E receptor 1, subtype EP1; prostaglandin E receptor EP1 subtype; prostaglandin E2 receptor EP1 subtype; Prostanoid EP1 receptor; PTGER1; Ptgerep1
Gene Symbols Ptger1
Host Species Rabbit
Immunogen Synthetic 16 amino acid peptide from 1st cytoplasmic domain of human PTGER1 / EP1.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5731
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.