Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PTGER4/EP4 Antibody, Novus Biologicals™

Goat Polyclonal Antibody has been used in 3 publications
Supplier: Novus Biologicals NB1002801
Description
PTGER4/EP4 Polyclonal specifically detects PTGER4/EP4 in Human, Mouse samples. It is validated for Western Blot, Peptide ELISA.Specifications
PTGER4/EP4 | |
Polyclonal | |
Unconjugated | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
EP4, EP4R, MGC126583, PGE receptor EP4 subtype, PGE receptor, EP4 subtype, PGE2 receptor EP4 subtype, prostaglandin E receptor 4 (subtype EP4), prostaglandin E2 receptor EP4 subtype, Prostanoid EP4 receptor, PTGER2 | |
Goat | |
Affinity Purified | |
RUO | |
Primary | |
Cross reacts with Human. Expected from sequence similarity: Dog. | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot, ELISA | |
0.5 mg/mL | |
Western Blot 0.1 - 0.3 ug/ml, Peptide ELISA Detection limit 1:16000 | |
NP_000949.1 | |
PTGER4 | |
Peptide with sequence C-LEREVSKNPDLQA corresponding to internal region according to NP_000949.1. | |
0.1 mg | |
Cancer | |
5734 | |
Human, Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction