Learn More
Invitrogen™ PTGES Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA533000
Description
Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey, Rabbit (100%); Marmoset, Dog, Elephant, Panda, Pike (92%); Bovine, Hamster, Pig, Xenopus (85%). For IHC(P), use heat induced antigen retrieval in pH 6.0 citrate buffer. After incubation with the primary antibody, slides were incubated with biotinylated secondary antibody, followed by alkaline phosphatase-streptavidin and chromogen.
PTGES is a glutathione-dependent prostaglandin E synthase. The expression of this gene has been shown to be induced by proinflammatory cytokine interleukin 1 beta (IL1B). Its expression can also be induced by tumor suppressor protein TP53, and may be involved in TP53 induced apoptosis.
Specifications
| PTGES | |
| Polyclonal | |
| Unconjugated | |
| PTGES | |
| 2410099E23Rik; D2Ertd369e; glutathione S-transferase 1-like 1; MGST1L1; MGST1-L1; MGST1-like 1; MGST-IV; Microsomal glutathione S-transferase 1-like 1; microsomal prostaglandin E synthase 1; microsomal prostaglandin E synthase-1; MPGES; MPGES1; mPGES-1; p53-induced apoptosis protein 12; p53-induced gene 12 protein; Pges; PIG12; PP102; PP1294; Prostaglandin E synthase; PTGES; TP53I12; tumor protein p53 inducible protein 12 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 100342171, 9536 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| O14684 | |
| PTGES | |
| Synthetic 13 amino acid peptide from 1st cytoplasmic domain of human PTGES. | |
| 50 μg | |
| Primary | |
| Human, Rabbit | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.