Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PTGES Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA534126
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA534126 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA534126 Supplier Invitrogen™ Supplier No. PA534126
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey, Marmoset, Mouse, Rat, Elephant, Rabbit, Pig (100%) Hamster, Bovine (94%).

PTGES is a glutathione-dependent prostaglandin E synthase. The expression of this gene has been shown to be induced by proinflammatory cytokine interleukin 1 beta (IL1B). Its expression can also be induced by tumor suppressor protein TP53, and may be involved in TP53 induced apoptosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PTGES
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene PTGES
Gene Accession No. O14684, Q9JM51
Gene Alias 2410099E23Rik; D2Ertd369e; glutathione S-transferase 1-like 1; MGST1L1; MGST1-L1; MGST1-like 1; MGST-IV; Microsomal glutathione S-transferase 1-like 1; microsomal prostaglandin E synthase 1; microsomal prostaglandin E synthase-1; MPGES; MPGES1; mPGES-1; p53-induced apoptosis protein 12; p53-induced gene 12 protein; Pges; PIG12; PP102; PP1294; Prostaglandin E synthase; PTGES; TP53I12; tumor protein p53 inducible protein 12
Gene Symbols PTGES
Host Species Rabbit
Immunogen Synthetic 16 amino acid peptide from N-Terminus of human PTGES.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100342171, 59103, 64292, 654407, 9536
Target Species Human, Mouse, Rat, Rabbit, Monkey, Pig
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.