Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PTGES Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560916
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560916 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560916 Supplier Invitrogen™ Supplier No. PA560916
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ITGQVRLRKK AFANPEDALR HGGPQYCRSD PDVERCLRAP RNDM Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 91%.

PTGES is a glutathione-dependent prostaglandin E synthase. The expression of this gene has been shown to be induced by proinflammatory cytokine interleukin 1 beta (IL1B). Its expression can also be induced by tumor suppressor protein TP53, and may be involved in TP53 induced apoptosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PTGES
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.20 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PTGES
Gene Accession No. O14684
Gene Alias 2410099E23Rik; D2Ertd369e; glutathione S-transferase 1-like 1; MGST1L1; MGST1-L1; MGST1-like 1; MGST-IV; Microsomal glutathione S-transferase 1-like 1; microsomal prostaglandin E synthase 1; microsomal prostaglandin E synthase-1; MPGES; MPGES1; mPGES-1; p53-induced apoptosis protein 12; p53-induced gene 12 protein; Pges; PIG12; PP102; PP1294; Prostaglandin E synthase; PTGES; TP53I12; tumor protein p53 inducible protein 12
Gene Symbols PTGES
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PTGES. Recombinant protein control fragment (Product #RP-100154).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9536
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.