Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PTGES3 Monoclonal Antibody (OTI2H10), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA803436

Description
Cytosolic prostaglandin synthase that catalyzes the oxidoreduction of prostaglandin endoperoxide H2 (PGH2) to prostaglandin E2 (PGE2). Molecular chaperone that localizes to genomic response elements in a hormone-dependent manner and disrupts receptor-mediated transcriptional activation, by promoting disassembly of transcriptional regulatory complexes. Facilitates HIF alpha proteins hydroxylation via interaction with EGLN1/PHD2, leading to recruit EGLN1/PHD2 to the HSP90 pathway.Specifications
PTGES3 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q15185 | |
PTGES3 | |
Full length human recombit protein of human PTGES3 produced in E.coli. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG2a |
Western Blot | |
OTI2H10 | |
Unconjugated | |
PTGES3 | |
P23, TEBP, cPGES | |
Mouse | |
Affinity Chromatography | |
RUO | |
10728 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction