Learn More
PTGES3 Mouse anti-Human, Clone: OTI2D2, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF803433

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Cytosolic prostaglandin synthase that catalyzes the oxidoreduction of prostaglandin endoperoxide H2 (PGH2) to prostaglandin E2 (PGE2). Molecular chaperone that localizes to genomic response elements in a hormone-dependent manner and disrupts receptor-mediated transcriptional activation, by promoting disassembly of transcriptional regulatory complexes. Facilitates HIF alpha proteins hydroxylation via interaction with EGLN1/PHD2, leading to recruit EGLN1/PHD2 to the HSP90 pathway.Specifications
PTGES3 | |
Monoclonal | |
Unconjugated | |
PTGES3 | |
P23, TEBP, cPGES | |
Mouse | |
Affinity Chromatography | |
RUO | |
10728 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
OTI2D2 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q15185 | |
PTGES3 | |
Full length human recombit protein of human PTGES3 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.