Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PTGFR Antibody (1014602) [FITC], Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB10294F
Description
PTGFR Monoclonal antibody specifically detects PTGFR in Human samples. It is validated for Flow Cytometry, CyTOFSpecifications
PTGFR | |
Monoclonal | |
FITC | |
PBS | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry, CyTOF | |
1014602 | |
Flow Cytometry, CyTOF-ready | |
FP, MGC120498, MGC46203, PGF receptor, PGF2 alpha receptor, PGF2-alpha receptor, prostaglandin F receptor (FP), prostaglandin F2 alpha receptor, prostaglandin F2-alpha receptor, prostaglandin receptor (2-alpha), Prostanoid FP receptor | |
Synthetic peptide containing human PTGFR RD3 epitope, Accession # P43088 | |
0.1 mL | |
GPCR | |
5737 | |
Store at 4°C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction