Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PTGFR Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA534130
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA534130 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA534130 Supplier Invitrogen™ Supplier No. PA534130
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey, Mouse, Elephant, Panda, Bat, Dog, Rabbit, Horse, Turkey, Chicken, Platypus (100%).

The protein encoded by this gene is member of the G-protein coupled receptor family. This protein is a receptor for prostaglandin F2-alpha (PGF2-alpha), which is known to be a potent luteolytic agent, and may also be involved in modulating intraocular pressure and smooth muscle contraction in uterus. Knockout studies in mice suggest that the interaction of PGF2-alpha with this receptor may initiate parturition in ovarian luteal cells and thus induce luteolysis. Two transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PTGFR
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene PTGFR
Gene Accession No. P43088, P43117
Gene Alias AI957154; fp; FP prostanoid receptor; FP prostanoid receptor FPB isoform; FP receptor; PF2AR; PGF; PGF receptor; PGF2 alpha receptor; PGF2-alpha receptor; prostaglandin F receptor; prostaglandin F receptor (FP); prostaglandin F2 alpha receptor; prostaglandin F2alpha receptor; prostaglandin F2-alpha receptor; prostaglandin F2alpha receptor FPa isoform; prostaglandin F2alpha receptor isoform-aplpha; prostaglandin F2alpha receptor isoform-beta; prostaglandin F2alpha receptor isoform-delta; prostaglandin F2alpha receptor isoform-epsiron; prostaglandin F2alpha receptor isoform-gamma; prostaglandin receptor (2-alpha); prostanoid FP receptor; Ptgfr
Gene Symbols PTGFR
Host Species Rabbit
Immunogen Synthetic 14 amino acid peptide from 1st cytoplasmic domain of human PTGFR.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100009714, 100344670, 19220, 424548, 479981, 5737
Target Species Human, Mouse, Canine, Rabbit, Horse, Chicken, Bat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.