Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PTGFR Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA570674
Description
This target displays homology in the following species: Cow: 92%; Dog: 100%; Guinea Pig: 86%; Horse: 93%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 93%; Sheep: 92%.
The protein encoded by this gene is member of the G-protein coupled receptor family. This protein is a receptor for prostaglandin F2-alpha (PGF2-alpha), which is known to be a potent luteolytic agent, and may also be involved in modulating intraocular pressure and smooth muscle contraction in uterus. Knockout studies in mice suggest that the interaction of PGF2-alpha with this receptor may initiate parturition in ovarian luteal cells and thus induce luteolysis. Two transcript variants encoding different isoforms have been found for this gene.
Specifications
PTGFR | |
Polyclonal | |
Unconjugated | |
PTGFR | |
AI957154; fp; FP prostanoid receptor; FP prostanoid receptor FPB isoform; FP receptor; PF2AR; PGF; PGF receptor; PGF2 alpha receptor; PGF2-alpha receptor; prostaglandin F receptor; prostaglandin F receptor (FP); prostaglandin F2 alpha receptor; prostaglandin F2alpha receptor; prostaglandin F2-alpha receptor; prostaglandin F2alpha receptor FPa isoform; prostaglandin F2alpha receptor isoform-aplpha; prostaglandin F2alpha receptor isoform-beta; prostaglandin F2alpha receptor isoform-delta; prostaglandin F2alpha receptor isoform-epsiron; prostaglandin F2alpha receptor isoform-gamma; prostaglandin receptor (2-alpha); prostanoid FP receptor; Ptgfr | |
Rabbit | |
Affinity chromatography | |
RUO | |
5737 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
P43088 | |
PTGFR | |
synthetic peptide directed towards the C-terminal region of human PTGFR. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction