Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PTGFRN Monoclonal Antibody (4E9G7)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA548581
Description
PTGFRN Monoclonal Antibody for Western Blot, IHC (P), Flow, ELISA
Inhibits the binding of prostaglandin F2-alpha (PGF2-alpha) to its specific FP receptor, by decreasing the receptor number rather than the affinity constant. Functional coupling with the prostaglandin F2-alpha receptor seems to occur. {ECO:0000250}.
Specifications
PTGFRN | |
Monoclonal | |
1 mg/mL | |
PBS with 0.05% sodium azide | |
Q9P2B2 | |
PTGFRN | |
Purified recombinant fragment of human CD315 (AA: extra 46-221) expressed in E. Coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |
ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Western Blot | |
4E9G7 | |
Unconjugated | |
PTGFRN | |
4833445A08Rik; AU042434; CD315; CD9 partner 1; CD9P1; CD9P-1; EWIF; EWI-F; FPRP; Glu-Trp-Ile EWI motif-containing protein F; KIAA1436; prostaglandin F2 receptor inhibitor; prostaglandin F2 receptor negative regulator; Prostaglandin F2-alpha receptor regulatory protein; prostaglandin F2-alpha receptor-associated protein; Ptgfrn; SMAP-6; Trim45 | |
Mouse | |
Protein G | |
RUO | |
5738 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction