Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PTGFRN Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA548054
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA548054 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA548054 Supplier Invitrogen™ Supplier No. PA548054
Add to Cart
Add to Cart

Sheep Polyclonal Antibody

Reconstitute at 0.2 mg/mL in sterile PBS.

Inhibits the binding of prostaglandin F2-alpha (PGF2-alpha) to its specific FP receptor, by decreasing the receptor number rather than the affinity constant. Functional coupling with the prostaglandin F2-alpha receptor seems to occur. {ECO:0000250}.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PTGFRN
Applications Flow Cytometry, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene PTGFRN
Gene Accession No. Q9WV91
Gene Alias 4833445A08Rik; AU042434; CD315; CD9 partner 1; CD9P1; CD9P-1; EWIF; EWI-F; FPRP; Glu-Trp-Ile EWI motif-containing protein F; KIAA1436; prostaglandin F2 receptor inhibitor; prostaglandin F2 receptor negative regulator; Prostaglandin F2-alpha receptor regulatory protein; prostaglandin F2-alpha receptor-associated protein; Ptgfrn; SMAP-6; Trim45
Gene Symbols PTGFRN
Host Species Sheep
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse FPRP Arg22-Pro832.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19221
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.