Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PTGFRN Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA551134
Description
PTGFRN / CD315 Polyclonal Antibody detects endogenous levels of PTGFRN / CD315 protein.
Inhibits the binding of prostaglandin F2-alpha (PGF2-alpha) to its specific FP receptor, by decreasing the receptor number rather than the affinity constant. Functional coupling with the prostaglandin F2-alpha receptor seems to occur. {ECO:0000250}.
Specifications
PTGFRN | |
Polyclonal | |
Unconjugated | |
PTGFRN | |
4833445A08Rik; AU042434; CD315; CD9 partner 1; CD9P1; CD9P-1; EWIF; EWI-F; FPRP; Glu-Trp-Ile EWI motif-containing protein F; KIAA1436; prostaglandin F2 receptor inhibitor; prostaglandin F2 receptor negative regulator; Prostaglandin F2-alpha receptor regulatory protein; prostaglandin F2-alpha receptor-associated protein; Ptgfrn; SMAP-6; Trim45 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
19221, 29602, 5738 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
1 mg/mL | |
PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q62786, Q9P2B2, Q9WV91 | |
PTGFRN | |
Synthesized peptide derived from the Internal region of human CD315 (aa 601-650). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction