Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PTGFRN Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553547
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553547 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553547 Supplier Invitrogen™ Supplier No. PA553547
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NSGYYYCHVS LWAPGHNRSW HKVAEAVSSP AGVGVTWLEP DYQVYLNASK VPGFADDPTE LACRVVDTKS GEANVRFTVS WYYRMNRRSD NVVTSELLAV MDGDW Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 82%.

Inhibits the binding of prostaglandin F2-alpha (PGF2-alpha) to its specific FP receptor, by decreasing the receptor number rather than the affinity constant. Functional coupling with the prostaglandin F2-alpha receptor seems to occur. {ECO:0000250}.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PTGFRN
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PTGFRN
Gene Accession No. Q9P2B2
Gene Alias 4833445A08Rik; AU042434; CD315; CD9 partner 1; CD9P1; CD9P-1; EWIF; EWI-F; FPRP; Glu-Trp-Ile EWI motif-containing protein F; KIAA1436; prostaglandin F2 receptor inhibitor; prostaglandin F2 receptor negative regulator; Prostaglandin F2-alpha receptor regulatory protein; prostaglandin F2-alpha receptor-associated protein; Ptgfrn; SMAP-6; Trim45
Gene Symbols PTGFRN
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PTGFRN. Recombinant protein control fragment (Product #RP-90234).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5738
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.