Learn More
Invitrogen™ PTGFRN Polyclonal Antibody

Description
Recommended positive controls: 293T, A431, H1299. Predicted reactivity: Mouse (100%), Rat (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | PTGFRN |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.8 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 20% glycerol and 0.025% ProClin 300; pH 7 |
| Gene | PTGFRN |
| Gene Accession No. | Q62786, Q9P2B2 |
| Gene Alias | 4833445A08Rik; AU042434; CD315; CD9 partner 1; CD9P1; CD9P-1; EWIF; EWI-F; FPRP; Glu-Trp-Ile EWI motif-containing protein F; KIAA1436; prostaglandin F2 receptor inhibitor; prostaglandin F2 receptor negative regulator; Prostaglandin F2-alpha receptor regulatory protein; prostaglandin F2-alpha receptor-associated protein; Ptgfrn; SMAP-6; Trim45 |
| Gene Symbols | PTGFRN |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.