Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PTGIR (extracellular) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA577695
Description
Product is shipped at room temperature as a lyophilized powder and should be stored at -20C upon receipt. Reconstitute using 50 uL deionized water.
Receptor for prostacyclin (prostaglandin I2 or PGI2). The activity of this receptor is mediated by G(s) proteins which activate adenylate cyclase.
Specifications
PTGIR (extracellular) | |
Polyclonal | |
Unconjugated | |
PTGIR | |
Prostanoid Prostacyclin Receptor (extracellular) | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
19222, 292661, 5739 | |
-20°C | |
Lyophilized |
Flow Cytometry, Immunofluorescence, Western Blot | |
0.8 mg/mL | |
PBS with 1% BSA and 0.05% sodium azide; pH 7.4 | |
P43119, P43252, P43253 | |
Ptgir | |
Peptide (C)TQAIAPDSREMGD, corresponding to amino acid residues 290-302 of rat Prostanoid Prostacyclin receptor | |
50 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction