Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PTGIR Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5102240
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5102240 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5102240 Supplier Invitrogen™ Supplier No. PA5102240
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total Prostacyclin Receptor.

Receptor for prostacyclin (prostaglandin I2 or PGI2). The activity of this receptor is mediated by G(s) proteins which activate adenylate cyclase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PTGIR
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene PTGIR
Gene Accession No. P43119
Gene Alias IP; IP receptor; PGI receptor; PGI2; PGI2 receptor; PRIPR; Prostacyclin receptor; prostaglandin I receptor (IP); prostaglandin I2 (prostacyclin) receptor (IP); prostaglandin I2 receptor; Prostanoid IP receptor; Ptgir
Gene Symbols PTGIR
Host Species Rabbit
Immunogen A synthesized peptide derived from human PTGIR(Accession P43119), corresponding to amino acid residues G204-P254.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5739
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.