Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PTGIS Polyclonal Antibody

Goat Polyclonal Antibody
Supplier: Invitrogen™ PA5143283
This item is not returnable.
View return policy
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:128,000.
CYP8A1 is a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. However, this protein is considered a member of the cytochrome P450 superfamily on the basis of sequence similarity rather than functional similarity. This endoplasmic reticulum membrane protein catalyzes the conversion of prostglandin H2 to prostacyclin (prostaglandin I2), a potent vasodilator and inhibitor of platelet aggregation. An imbalance of prostacyclin and its physiological antagonist thromboxane A2 contribute to the development of myocardial infarction, stroke, and atherosclerosis.
Specifications
PTGIS | |
Polyclonal | |
Unconjugated | |
PTGIS | |
Cyp8; CYP8A1; cytochrome P450 8A1; cytochrome P450, family 8, subfamily A, polypeptide 1; hydroperoxy icosatetraenoate dehydratase; LOW QUALITY PROTEIN: prostacyclin synthase; MGC126858; MGC126860; PGIS; Prostacyclin synthase; prostaglandin I2 (prostacyclin) synthase; prostaglandin I2 (prostacyclin) synthase like; prostaglandin I2 synthase; PTGI; ptgis; ptgisl | |
Goat | |
Ammonium sulfate precipitation | |
RUO | |
5740 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q16647 | |
PTGIS | |
Peptide with sequence C-RRAHRSKWLES. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction