Learn More
Invitrogen™ PTGR1 Polyclonal Antibody

Description
PTGR1 Polyclonal Antibody for Western Blot
Specifications
Specifications
| Antigen | PTGR1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | PTGR1 |
| Gene Accession No. | Q14914, Q91YR9 |
| Gene Alias | 12-HD; 15-oxoprostaglandin 13-reductase; 2510002C21Rik; ADRAB-F; D3T-inducible gene 1 protein; Dig1; DIG-1; Dithiolethione-inducible gene 1 protein; dithiolethione-inducible gene-1; FLJ99229; leukotriene B4 12-hydroxydehydrogenase; leukotriene b4 12-hydroxydehydrogenase/prostaglandin 15-keto reductase; LTB4; LTB4DH; MGC34943; NADP-dependent leukotriene B4 12-hydroxydehydrogenase; PGR; PGR1; PRG-1; Prostaglandin reductase 1; PTGR1; unnamed protein product; Zadh3; zinc binding alcohol dehydrogenase domain containing 3 |
| Gene Symbols | PTGR1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.