Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PTGR1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA544230
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA544230 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA544230 Supplier Invitrogen™ Supplier No. PA544230
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: VRTKTWTLKK HFVGYPTNSD FELKTSELPP LKNGEVLLEA LFLTVDPYMR Sequence homology: Cow: 93%; Guinea Pig: 83%; Horse: 83%; Human: 93%; Rabbit: 83%; Rat: 79%.

PTGR1 encodes an enzyme that is involved in the inactivation of the chemotactic factor, leukotriene B4. The encoded protein specifically catalyzes the NADP+ dependent conversion of leukotriene B4 to 12-oxo-leukotriene B4. A pseudogene of this gene is found on chromosome 1. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PTGR1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene PTGR1
Gene Accession No. Q14914
Gene Alias 12-HD; 15-oxoprostaglandin 13-reductase; 2510002C21Rik; ADRAB-F; D3T-inducible gene 1 protein; Dig1; DIG-1; Dithiolethione-inducible gene 1 protein; dithiolethione-inducible gene-1; FLJ99229; leukotriene B4 12-hydroxydehydrogenase; leukotriene b4 12-hydroxydehydrogenase/prostaglandin 15-keto reductase; LTB4; LTB4DH; MGC34943; NADP-dependent leukotriene B4 12-hydroxydehydrogenase; PGR; PGR1; PRG-1; Prostaglandin reductase 1; PTGR1; unnamed protein product; Zadh3; zinc binding alcohol dehydrogenase domain containing 3
Gene Symbols PTGR1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human LTB4DH.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22949
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.