Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PTGR1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 133741AP150UL
Description
PTGR1 encodes an enzyme that is involved in the inactivation of the chemotactic factor, leukotriene B4. The encoded protein specifically catalyzes the NADP+ dependent conversion of leukotriene B4 to 12-oxo-leukotriene B4. A pseudogene of this gene is found on chromosome 1. Alternative splicing results in multiple transcript variants.Specifications
| PTGR1 | |
| Polyclonal | |
| Unconjugated | |
| PTGR1 | |
| LTB4DH, PGR1, PRG 1, prostaglandin reductase 1, PTGR1, ZADH3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 192227, 22949, 67103 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P97584, Q14914, Q91YR9 | |
| Ptgr1 | |
| PTGR1 Fusion Protein Ag4196 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction