Learn More
PTGS2 Mouse anti-Human, Clone: OTI10H5, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF805307

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Prostaglandin-endoperoxide synthase (PTGS), also known as cyclooxygenase, is the key enzyme in prostaglandin biosynthesis, and acts both as a dioxygenase and as a peroxidase. There are two isozymes of PTGS: a constitutive PTGS1 and an inducible PTGS2, which differ in their regulation of expression and tissue distribution. This gene encodes the inducible isozyme. It is regulated by specific stimulatory events, suggesting that it is responsible for the prostanoid biosynthesis involved in inflammation and mitogenesis.Specifications
PTGS2 | |
Monoclonal | |
Unconjugated | |
PTGS2 | |
COX-2, COX2, GRIPGHS, PGG/HS, PGHS-2, PHS-2, hCox-2 | |
Mouse | |
Affinity Chromatography | |
RUO | |
5743 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
OTI10H5 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P35354 | |
PTGS2 | |
Human recombit protein fragment corresponding to amino acids 18-260 of human PTGS2 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.