Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PTH Antibody (PTH/1717R) - N-terminal, Novus Biologicals™
SDP

Rabbit Monoclonal Antibody

$304.50 - $560.50

Specifications

Antigen PTH
Clone PTH/1717R
Concentration 0.2 mg/mL
Dilution Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 0.5 - 1 ug/ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB25317020
SDP
View Documents
Novus Biologicals
NBP25317020UG
20 μg
Each for $304.50
Only null left
Add to Cart
 
NB253170100
SDP
View Documents
Novus Biologicals
NBP253170100UG
100 μg
Each for $560.50
Only null left
Add to Cart
 
NB253170200
SDP
View Documents
Novus Biologicals
NBP253170200UG
200 μg N/A N/A N/A
Description

Description

PTH Monoclonal specifically detects PTH in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

PTH
0.2 mg/mL
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Apoptosis, Biologically Active Proteins, Cancer
10mM PBS with 0.05% BSA with 0.05% Sodium Azide
5741
Recombinant human PTH protein N-terminal fragment (exact sequence is proprietary)
Primary
Store at 4C.
PTH/1717R
Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 0.5 - 1 ug/ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml
Monoclonal
Purified
RUO
Human
Parathormone, Parathyrin, parathyroid hormone, parathyroid hormone 1, PTH1
PTH
IgG κ
Protein A or G purified
9 kDa
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.